Assay Details
Gene Symbol: fen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328507.1 | NP_001315436.1 | 10 - 11 | 1080 | 100 | |
XM_017353804.1 | XP_017209293.1 | 10 - 11 | 1098 | 100 | ||
GenBank mRNA | AY391423.1 | - | 10 - 11 | 1009 | 100 | |
AY648844.1 | - | 10 - 11 | 1022 | 100 | ||
BC049413.1 | - | 10 - 11 | 1297 | 100 | ||
BC071488.1 | - | 10 - 11 | 1041 | 100 | ||
CT678576.2 | - | 2 - 3 | 363 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
