Assay Details
Gene Symbol: vamp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200005.1 | NP_956299.1 | 2 - 3 | 330 | 71 | |
GenBank mRNA | BC059626.1 | - | 2 - 3 | 330 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
