Assay Details
Gene Symbol: cdc42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001018120.2 | NP_001018130.1 | 2 - 3 | 242 | 109 | |
NM_200632.2 | NP_956926.1 | 2 - 3 | 242 | 109 | ||
GenBank mRNA | AY398322.1 | - | 2 - 3 | 235 | 109 | |
AY865566.1 | - | 1 - 2 | 107 | 109 | ||
BC057415.1 | - | 1 - 2 | 143 | 109 | ||
EH509759.1 | - | 2 - 3 | 222 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
