Assay Details
Gene Symbol: cdc23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200933.1 | NP_957227.1 | 7 - 8 | 819 | 85 | |
GenBank mRNA | BC045861.1 | - | 7 - 8 | 819 | 85 | |
BC155283.1 | - | 7 - 8 | 783 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
