Assay Details
Gene Symbol: cdc23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200933.1 | NP_957227.1 | 8 - 9 | 925 | 84 | |
GenBank mRNA | BC045861.1 | - | 8 - 9 | 925 | 84 | |
BC155283.1 | - | 8 - 9 | 889 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
