Assay Details
Gene Symbol: clic4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201486.1 | NP_958894.1 | 4 - 5 | 699 | 106 | |
GenBank mRNA | AY398328.1 | - | 4 - 5 | 679 | 106 | |
BC051622.1 | - | 4 - 5 | 699 | 106 | ||
BC065609.1 | - | 4 - 5 | 614 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
