Assay Details
Gene Symbol: ube2r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002600.1 | NP_001002600.1 | 4 - 5 | 553 | 78 | |
XM_005161018.4 | XP_005161075.1 | 4 - 5 | 1102 | 78 | ||
GenBank mRNA | BC075995.1 | - | 4 - 5 | 553 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
