Assay Details
Gene Symbol: crhbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003459.1 | NP_001003459.1 | 1 - 2 | 118 | 118 | |
GenBank mRNA | BC076472.1 | - | 1 - 2 | 118 | 118 | |
BC164122.1 | - | 1 - 2 | 114 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
