Assay Details
Gene Symbol: hsd17b14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003521.2 | NP_001003521.1 | 1 - 2 | 206 | 58 | |
GenBank mRNA | BC077136.1 | - | 1 - 2 | 210 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
