Assay Details
Gene Symbol: rras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005931.2 | NP_001005931.2 | 5 - 6 | 688 | 100 | |
GenBank mRNA | BC096943.1 | - | 5 - 6 | 688 | 100 | |
BC163945.1 | - | 4 - 5 | 418 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
