Assay Details
Gene Symbol: abraxas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005993.1 | NP_001005993.1 | 7 - 8 | 760 | 113 | |
XM_021468815.1 | XP_021324490.1 | 7 - 8 | 889 | 113 | ||
XM_021468816.1 | XP_021324491.1 | 7 - 8 | 768 | 113 | ||
GenBank mRNA | BC083413.1 | - | 7 - 8 | 760 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
