Assay Details
Gene Symbol: slc7a6os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007344.2 | NP_001007345.2 | 2 - 3 | 673 | 87 | |
GenBank mRNA | AB193555.1 | - | 1 - 2 | 977 | 87 | |
BC085534.1 | - | 2 - 3 | 652 | 87 | ||
BC165325.1 | - | 2 - 3 | 550 | 87 | ||
EH556070.1 | - | 2 - 3 | 648 | 87 | ||
EH577171.1 | - | 3 - 4 | 846 | 87 | ||
EH580278.1 | - | 2 - 3 | 683 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
