Assay Details
Gene Symbol: slc7a6os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007344.2 | NP_001007345.2 | 3 - 4 | 879 | 56 | |
GenBank mRNA | AB193555.1 | - | 2 - 3 | 1183 | 56 | |
BC085534.1 | - | 3 - 4 | 858 | 56 | ||
BC165325.1 | - | 3 - 4 | 756 | 56 | ||
EH577171.1 | - | 2 - 3 | 640 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
