Assay Details
Gene Symbol: amh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007779.1 | NP_001007780.1 | 1 - 2 | 113 | 82 | |
XM_009296234.3 | XP_009294509.1 | 1 - 2 | 113 | 82 | ||
GenBank mRNA | AY677080.2 | - | 1 - 2 | 196 | 82 | |
AY721604.1 | - | 1 - 2 | 113 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
