Assay Details
Gene Symbol: amh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007779.1 | NP_001007780.1 | 3 - 4 | 560 | 118 | |
XM_009296234.3 | XP_009294509.1 | 3 - 4 | 118 | |||
GenBank mRNA | AY677080.2 | - | 3 - 4 | 643 | 118 | |
AY721604.1 | - | 3 - 4 | 560 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
