Assay Details
Gene Symbol: fn1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013261.1 | NP_001013279.1 | 22 - 23 | 3585 | 69 | |
XM_005160284.3 | XP_005160341.1 | 22 - 23 | 3620 | 69 | ||
XM_009305554.3 | XP_009303829.1 | 22 - 23 | 3621 | 69 | ||
GenBank mRNA | AY538257.1 | - | 22 - 23 | 3585 | 69 | |
AY725818.1 | - | 22 - 23 | 3600 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
