Assay Details
Gene Symbol: fn1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013261.1 | NP_001013279.1 | 23 - 24 | 3673 | 72 | |
XM_005160284.3 | XP_005160341.1 | 23 - 24 | 3708 | 72 | ||
XM_009305554.3 | XP_009303829.1 | 23 - 24 | 3709 | 72 | ||
GenBank mRNA | AY538257.1 | - | 23 - 24 | 3673 | 72 | |
AY725818.1 | - | 23 - 24 | 3688 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
