Assay Details
Gene Symbol: rgs5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017821.2 | NP_001017821.1 | 1 - 2 | 124 | 68 | |
GenBank mRNA | BC092795.1 | - | 1 - 2 | 125 | 68 | |
BC165301.1 | - | 1 - 2 | 78 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
