Assay Details
Gene Symbol: otofa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030112.2 | NP_001025283.1 | 11 - 12 | 1061 | 157 | |
XM_009294732.3 | XP_009293007.1 | 11 - 12 | 1360 | 157 | ||
XM_009294733.3 | XP_009293008.1 | 11 - 12 | 1361 | 157 | ||
XM_009294734.3 | XP_009293009.1 | 11 - 12 | 1360 | 157 | ||
XM_009294736.3 | XP_009293011.1 | 11 - 12 | 1360 | 157 | ||
XM_009294738.3 | XP_009293013.1 | 11 - 12 | 1361 | 157 | ||
XM_009294739.3 | XP_009293014.1 | 11 - 12 | 1360 | 157 | ||
XM_009294740.3 | XP_009293015.1 | 11 - 12 | 1360 | 157 | ||
XM_009294742.3 | XP_009293017.1 | 11 - 12 | 1360 | 157 | ||
XM_009294744.3 | XP_009293019.1 | 11 - 12 | 1360 | 157 | ||
XM_021468441.1 | XP_021324116.1 | 11 - 12 | 1360 | 157 | ||
XM_021468442.1 | XP_021324117.1 | 11 - 12 | 1360 | 157 | ||
XM_021468443.1 | XP_021324118.1 | 11 - 12 | 1360 | 157 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
