Assay Details
Gene Symbol: dscama | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030224.2 | NP_001025395.1 | 21 - 22 | 4427 | 112 | |
XM_005155472.3 | XP_005155529.1 | 15 - 16 | 2501 | 112 | ||
XM_021479191.1 | XP_021334866.1 | 21 - 22 | 4504 | 112 | ||
GenBank mRNA | AY552547.1 | - | 21 - 22 | 4427 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
