Assay Details
Gene Symbol: tnikb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037676.2 | NP_001032765.2 | 26 - 27 | 3359 | 100 | |
XM_005162778.4 | XP_005162835.1 | 26 - 27 | 3719 | 100 | ||
XM_005162779.4 | XP_005162836.1 | 25 - 26 | 3364 | 100 | ||
XM_005162780.4 | XP_005162837.1 | 24 - 25 | 3277 | 100 | ||
XM_021470183.1 | XP_021325858.1 | 28 - 29 | 3752 | 100 | ||
XM_021470184.1 | XP_021325859.1 | 27 - 28 | 3665 | 100 | ||
XM_021470185.1 | XP_021325860.1 | 27 - 28 | 3605 | 100 | ||
XM_021470186.1 | XP_021325861.1 | 28 - 29 | 3753 | 100 | ||
XM_021470187.1 | XP_021325862.1 | 27 - 28 | 3397 | 100 | ||
XM_021470188.1 | XP_021325863.1 | 26 - 27 | 3310 | 100 | ||
GenBank mRNA | BC108020.1 | - | 7 - 8 | 974 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
