Assay Details
Gene Symbol: tnikb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037676.2 | NP_001032765.2 | 27 - 28 | 3504 | 105 | |
XM_005162778.4 | XP_005162835.1 | 27 - 28 | 3864 | 105 | ||
XM_005162779.4 | XP_005162836.1 | 26 - 27 | 3509 | 105 | ||
XM_005162780.4 | XP_005162837.1 | 25 - 26 | 3422 | 105 | ||
XM_021470183.1 | XP_021325858.1 | 29 - 30 | 3897 | 105 | ||
XM_021470184.1 | XP_021325859.1 | 28 - 29 | 3810 | 105 | ||
XM_021470185.1 | XP_021325860.1 | 28 - 29 | 3750 | 105 | ||
XM_021470186.1 | XP_021325861.1 | 29 - 30 | 3898 | 105 | ||
XM_021470187.1 | XP_021325862.1 | 28 - 29 | 3542 | 105 | ||
XM_021470188.1 | XP_021325863.1 | 27 - 28 | 3455 | 105 | ||
GenBank mRNA | BC108020.1 | - | 8 - 9 | 1119 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
