Assay Details
Gene Symbol: srd5a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044939.1 | NP_001038404.1 | 2 - 3 | 385 | 79 | |
GenBank mRNA | AW280458.1 | - | 2 - 3 | 375 | 79 | |
BC097226.1 | - | 2 - 3 | 376 | 79 | ||
BC117626.1 | - | 2 - 3 | 396 | 79 | ||
BC133964.1 | - | 2 - 3 | 385 | 79 | ||
CB353112.1 | - | 2 - 3 | 392 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
