Assay Details
Gene Symbol: srd5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076653.1 | NP_001070121.1 | 5 - 6 | 895 | 88 | |
GenBank mRNA | BC124451.1 | - | 5 - 6 | 895 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
