Assay Details
Gene Symbol: mettl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077342.1 | NP_001070810.1 | 4 - 5 | 664 | 77 | |
GenBank mRNA | BC124631.1 | - | 4 - 5 | 664 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
