Assay Details
Gene Symbol: taf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079973.1 | NP_001073442.1 | 11 - 12 | 906 | 79 | |
XM_005173352.4 | XP_005173409.1 | 11 - 12 | 911 | 79 | ||
XM_021481420.1 | XP_021337095.1 | 10 - 11 | 917 | 79 | ||
XM_021481421.1 | XP_021337096.1 | 10 - 11 | 860 | 79 | ||
GenBank mRNA | BC128851.1 | - | 11 - 12 | 906 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
