Assay Details
Gene Symbol: taf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079973.1 | NP_001073442.1 | 13 - 14 | 1118 | 143 | |
XM_005173352.4 | XP_005173409.1 | 13 - 14 | 1123 | 143 | ||
XM_021481420.1 | XP_021337095.1 | 12 - 13 | 1129 | 143 | ||
XM_021481421.1 | XP_021337096.1 | 12 - 13 | 1072 | 143 | ||
GenBank mRNA | BC128851.1 | - | 13 - 14 | 1118 | 143 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
