Assay Details
Gene Symbol: rnasel3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099453.1 | NP_001092923.1 | 3 - 4 | 846 | 106 | |
GenBank mRNA | EF382669.1 | - | 3 - 4 | 846 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
