Assay Details
Gene Symbol: stxbp5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099981.2 | NP_001093451.1 | 8 - 9 | 826 | 74 | |
XM_009294727.3 | XP_009293002.1 | 8 - 9 | 1245 | 74 | ||
XM_009294728.3 | XP_009293003.1 | 8 - 9 | 1245 | 74 | ||
XM_009294730.3 | XP_009293005.1 | 8 - 9 | 1245 | 74 | ||
XM_021468429.1 | XP_021324104.1 | 8 - 9 | 1245 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
