Assay Details
Gene Symbol: rangrf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099995.1 | NP_001093465.1 | 3 - 4 | 346 | 78 | |
XM_005165494.4 | XP_005165551.1 | 4 - 5 | 724 | 78 | ||
XM_005165495.2 | XP_005165552.1 | 4 - 5 | 425 | 78 | ||
XM_009301822.3 | XP_009300097.1 | 4 - 5 | 526 | 78 | ||
GenBank mRNA | BC095195.1 | - | 4 - 5 | 674 | 78 | |
BC122253.1 | - | 4 - 5 | 681 | 78 | ||
BC133096.1 | - | 3 - 4 | 794 | 78 | ||
BC150378.1 | - | 3 - 4 | 839 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
