Assay Details
Gene Symbol: mest | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328532.1 | NP_001315461.1 | 11 - 12 | 1079 | 86 | |
NM_131043.2 | NP_571118.2 | 11 - 12 | 1082 | 86 | ||
GenBank mRNA | AF194334.1 | - | 11 - 12 | 985 | 86 | |
BC059471.1 | - | 11 - 12 | 1005 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
