Assay Details
Gene Symbol: mest | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328532.1 | NP_001315461.1 | 2 - 3 | 378 | 90 | |
NM_131043.2 | NP_571118.2 | 2 - 3 | 381 | 90 | ||
GenBank mRNA | AF194334.1 | - | 2 - 3 | 284 | 90 | |
BC059471.1 | - | 2 - 3 | 304 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
