Assay Details
Gene Symbol: mest | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328532.1 | NP_001315461.1 | 3 - 4 | 452 | 93 | |
NM_131043.2 | NP_571118.2 | 3 - 4 | 455 | 93 | ||
GenBank mRNA | AF194334.1 | - | 3 - 4 | 358 | 93 | |
BC059471.1 | - | 3 - 4 | 378 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
