Assay Details
Gene Symbol: tnfrsfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131840.2 | NP_571915.2 | 3 - 4 | 554 | 77 | |
GenBank mRNA | AF250042.2 | - | 3 - 4 | 77 | ||
BC162652.1 | - | 3 - 4 | 77 | |||
DQ812116.1 | - | 3 - 4 | 319 | 77 | ||
EB956321.1 | - | 3 - 4 | 509 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
