Assay Details
Gene Symbol: tnfrsfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131840.2 | NP_571915.2 | 9 - 10 | 1253 | 104 | |
GenBank mRNA | AF250042.2 | - | 9 - 10 | 1189 | 105 | |
BC162652.1 | - | 9 - 10 | 1153 | 104 | ||
DQ812116.1 | - | 9 - 10 | 1018 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
