Assay Details
Gene Symbol: mrpl45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328585.1 | NP_001315514.1 | 7 - 8 | 902 | 74 | |
NM_200307.2 | NP_956601.2 | 7 - 8 | 789 | 74 | ||
GenBank mRNA | BC050166.1 | - | 7 - 8 | 776 | 74 | |
EH576059.1 | - | 1 - 2 | 315 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
