Assay Details
Gene Symbol: snrpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205667.1 | NP_991230.1 | 4 - 5 | 514 | 129 | |
GenBank mRNA | AF259079.1 | - | 4 - 5 | 484 | 129 | |
BC054570.1 | - | 4 - 5 | 506 | 129 | ||
BC065601.1 | - | 4 - 5 | 514 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
