Assay Details
Gene Symbol: casc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205716.1 | NP_991279.1 | 6 - 7 | 797 | 92 | |
GenBank mRNA | AJ555546.1 | - | 6 - 7 | 797 | 92 | |
BC117611.1 | - | 6 - 7 | 796 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
