Assay Details
Gene Symbol: traf4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205762.1 | NP_991325.1 | 5 - 6 | 873 | 70 | |
GenBank mRNA | AJ556548.1 | - | 5 - 6 | 936 | 70 | |
BC065969.1 | - | 5 - 6 | 873 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
