Assay Details
Gene Symbol: relt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207073.1 | NP_996956.1 | 3 - 4 | 271 | 66 | |
GenBank mRNA | BC066389.1 | - | 3 - 4 | 271 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
