Assay Details
Gene Symbol: rcan1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003616.1 | NP_001003616.1 | 3 - 4 | 723 | 106 | |
XM_005167827.4 | XP_005167884.1 | 3 - 4 | 783 | 106 | ||
GenBank mRNA | BC078255.1 | - | 3 - 4 | 723 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
