Assay Details
Gene Symbol: flvcr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025522.1 | NP_001020693.1 | 5 - 6 | 1110 | 93 | |
XM_005160739.3 | XP_005160796.1 | 5 - 6 | 1537 | 93 | ||
XM_009294763.3 | XP_009293038.1 | 6 - 7 | 1418 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
