Assay Details
Gene Symbol: srp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033741.1 | NP_001028913.1 | 3 - 4 | 279 | 84 | |
GenBank mRNA | BC096882.1 | - | 3 - 4 | 279 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
