Assay Details
Gene Symbol: rell2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040301.1 | NP_001035391.1 | 4 - 5 | 591 | 68 | |
XM_005157154.4 | XP_005157211.1 | 5 - 6 | 700 | 68 | ||
GenBank mRNA | BC115295.1 | - | 4 - 5 | 591 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
