Assay Details
Gene Symbol: shoc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044786.1 | NP_001038251.1 | 3 - 4 | 1224 | 126 | |
XM_005161796.4 | XP_005161853.1 | 3 - 4 | 1225 | 126 | ||
GenBank mRNA | BC155579.1 | - | 3 - 4 | 1200 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
