Assay Details
Gene Symbol: shoc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044786.1 | NP_001038251.1 | 5 - 6 | 1546 | 84 | |
XM_005161796.4 | XP_005161853.1 | 5 - 6 | 1547 | 84 | ||
GenBank mRNA | BC155579.1 | - | 5 - 6 | 1522 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
