Assay Details
Gene Symbol: shoc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044786.1 | NP_001038251.1 | 6 - 7 | 1662 | 145 | |
XM_005161796.4 | XP_005161853.1 | 6 - 7 | 1663 | 145 | ||
GenBank mRNA | BC155579.1 | - | 6 - 7 | 1638 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
