Assay Details
Gene Symbol: tnfsf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076607.1 | NP_001070075.1 | 3 - 4 | 632 | 104 | |
GenBank mRNA | BC124495.1 | - | 3 - 4 | 632 | 104 | |
BC154255.1 | - | 3 - 4 | 320 | 104 | ||
HM439092.1 | - | 3 - 4 | 303 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
