Assay Details
Gene Symbol: tnfsf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076607.1 | NP_001070075.1 | 4 - 5 | 659 | 91 | |
GenBank mRNA | BC124495.1 | - | 4 - 5 | 659 | 91 | |
BC154255.1 | - | 4 - 5 | 347 | 91 | ||
HM439092.1 | - | 4 - 5 | 330 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
