Assay Details
Gene Symbol: rhebl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076748.2 | NP_001070216.1 | 7 - 8 | 774 | 99 | |
GenBank mRNA | BC124281.1 | - | 7 - 8 | 680 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
